Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodopropane, 98+%, stab. with copper

Catalog No. p-7021871
Click to view available options
Quantity:
100 g
500 g

CAS: 107-08-4 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
SMILES CC(C)I

Specifications

Melting Point °C to 101°C
Density 1.744
Boiling Point 101°C to 102°C
Flash Point 44°C (111°F)
Assay Percent Range ≥98%, Stabilized with copper
Refractive Index 1.505
Linear Formula CH3CH2CH2I
Quantity 100 g
UN Number UN2392
Beilstein 505937
Sensitivity Light sensitive
Merck Index 14,7862
Solubility Information Fully miscible in water.
IUPAC Name 1-iodopropane
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, stabilized with copper
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: IODOPROPANES
EINECSNumber 203-460-2
RTECSNumber TZ4100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.