Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodopropane, 99%, stabilized

Catalog No. AC174050010
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 107-08-4 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
IUPAC Name 1-iodopropane
SMILES CC(C)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101.0°C
Color Colorless to Yellow
Density 1.7400g/mL
Boiling Point 101.0°C to 102.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 1.5% max. (Sum of all by-products) (GC)
Packaging Glass bottle
Refractive Index 1.5030 to 1.5050
Linear Formula CH3CH2CH2I
Quantity 1 L
Beilstein 01, 113
Specific Gravity 1.74
Merck Index 15, 7974
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 169.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 203-460-2
RTECSNumber TZ4100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.