Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isochromanone 98.0+%, TCI America™
SDP

Catalog No. I09531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09531G 1 g
I09535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09531G Supplier TCI America Supplier No. I09531G

Chemical Identifiers

CAS 4702-34-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00799232
InChI Key XVTAQSGZOGYIEY-UHFFFAOYSA-N
Synonym 3,4-Dihydro-1H-2-benzopyran-1-one
PubChem CID 78429
ChEBI CHEBI:23745
IUPAC Name 3,4-dihydro-1H-2-benzopyran-1-one
SMILES O=C1OCCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 160°C
Quantity 1 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Isochromanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.