Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropyl-4-piperidone, 99%

Catalog No. p-7081480
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 5355-68-0 | C8H15NO | 141.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5355-68-0
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00038035
InChI Key CCDBCHAQIXKJCG-UHFFFAOYSA-N
Synonym 1-isopropyl-4-piperidone, 1-isopropylpiperidin-4-one, n-isopropyl-4-piperidone, 4-piperidinone, 1-1-methylethyl, 1-isopropyl-4-piperidinone, 4-piperidone, 1-isopropyl, 1-propan-2-yl piperidin-4-one, 1-1'-methylethyl-4-piperidone, 1-methylethyl piperidin-4-one
PubChem CID 79313
IUPAC Name 1-propan-2-ylpiperidin-4-one
SMILES CC(C)N1CCC(=O)CC1

Specifications

Density 0.95
Boiling Point 100°C to 101°C (27 mmHg)
Refractive Index 1.4655
Quantity 10 g
Beilstein 107857
Formula Weight 141.22
Percent Purity 99%
Chemical Name or Material 1-Isopropyl-4-piperidone
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 226-339-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.