Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AC430560010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454482-11-2
Molecular Formula C12H22BNO2
Molecular Weight (g/mol) 223.13
MDL Number MFCD11506069
InChI Key SQMVRFXDBRYXFQ-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine, 1-methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine, 1-methyl-1,2,3,6-tetrahydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, 4,4,5,5-tetramethyl-2-1-methyl-3,5-dihydro-2h-pyrid-4-yl-1,3,2-dioxaborolane, 1-methyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester, 1-methyl-1,2,3,6-tetrahydropyridin-4-yl boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1,2,3,6-tetrahydro-pyridine
PubChem CID 20773371
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.2°C to 75.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 223.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause long lasting harmful eff
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.