Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1,4-cyclohexadiene, 97%, stab. with 0.01% BHT

Catalog No. aab2029814
Click to view available options
Quantity:
25 g

CAS: 4313-57-9 | C7H10 | 94.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4313-57-9
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00001538
InChI Key QDXQAOGNBCOEQX-UHFFFAOYSA-N
Synonym 1-methyl-1,4-cyclohexadiene, 2,5-dihydrotoluene, 1,4-cyclohexadiene, 1-methyl, 1, 1-methyl, acmc-1ast4, 1-methyl1,4-cyclohexadiene, 1-methylcyclohexane-1,4-diene, 1-methyl-1,4-cyclohexadiene, stabilized
PubChem CID 78006
IUPAC Name 1-methylcyclohexa-1,4-diene
SMILES CC1=CCC=CC1

Specifications

Density 0.838
Boiling Point 114°C to 115°C
Flash Point 10°C (50°F)
Odor Characteristic
Refractive Index 1.471
Quantity 25 g
UN Number UN3295
Beilstein 1560168
Formula Weight 94.16
Percent Purity 97%
Chemical Name or Material 1-Methyl-1,4-cyclohexadiene, stabilized with 0.01% BHT
Show More Show Less
Hazard Category H225-H304-H319
Hazard Statement GHS H Statement
H225-H304-H319
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 224-329-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.