Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-phenylhydrazine, 96%

Catalog No. p-166315
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 618-40-6 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-40-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007627
InChI Key MWOODERJGVWYJE-UHFFFAOYSA-N
Synonym 1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
PubChem CID 36299
IUPAC Name 1-methyl-1-phenylhydrazine
SMILES CN(C1=CC=CC=C1)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 75°C to 78°C (0.4 kPa)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5810 to 1.585
Linear Formula C6H5N(CH3)NH2
Quantity 5 g
Beilstein 15, 117
Specific Gravity 1.04
Solubility Information Solubility in water: sparingly soluble in water.,miscible in ethanol,ether,benzene,chloroform
Formula Weight 122.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-phenylhydrazine, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-547-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.