Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-1-phenylhydrazine, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1-Methyl-1-phenylhydrazine is used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methyl-1-phenylhydrazine is used to make other chemicals.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.
1-Methyl-1-phenylhydrazine is used to make other chemicals.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 1.038 |
Boiling Point | 54°C to 55°C (0.3 mmHg) |
Flash Point | 92°C (198°F) |
Refractive Index | 1.583 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 122.17 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1-phenylhydrazine |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335-H227 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 210-547-9 |
RTECSNumber | MV7712000 |
TSCA | Yes |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction