Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 7.68, 90.62 |
CAS Max % | 9.38, 92.32 |
Color | Colorless to Yellow |
Density | 0.9340g/mL |
Boiling Point | 67.0°C |
Flash Point | −26°C |
Packaging | AcroSeal™ Glass Bottle |
Quantity | 100 mL |
Specific Gravity | 0.934 |
Solubility Information | Solubility in water: reacts |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. In contact with water releases flammable gases. Reacts violently with water. May form explo |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a dry place. Store in a closed conta |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction