Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-1H-benzimidazol-2-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC03813DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 20028-40-4
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H11N3
Quantity 1g
Synonym 1-methyl-1h-benzimidazol-2-yl methylamine, 1-methyl-1h-benzo d imidazol-2-yl methanamine, 1h-benzimidazole-2-methanamine, 1-methyl, c-1-methyl-1h-benzoimidazol-2-yl-methylamine, 1-methyl-1h-1,3-benzodiazol-2-yl methanamine, 1-1-methyl-1h-benzimidazol-2-yl methanamine, 2-aminomethyl-1-methylbenzimidazole, 1-methylbenzimidazol-2-yl methylamine, r-methyl-1h-benzimidazole-2-methanamine, s-methyl-1h-benzimidazole-2-methanamine
InChI Key GFQZSGGPNZDNBC-UHFFFAOYSA-N
SMILES CN1C2=CC=CC=C2N=C1CN
IUPAC Name (1-methylbenzimidazol-2-yl)methanamine
Molecular Weight (g/mol) 161.208
PubChem CID 800907
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-benzimidazol-2-yl)methylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.