Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

Catalog No. CC03609CB
Click to view available options
:
Amber Glass Bottle; 250mg

Specifications

CAS 17289-25-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H8N2O
Quantity 250mg
MDL Number MFCD06738905
Synonym 1-methyl-1h-imidazol-4-yl methanol, 1-methylimidazol-4-yl methanol, 1-methyl-4-imidazolemethanol, 1-methylimidazole-4-methanol, 1h-imidazole-4-methanol,1-methyl, 1h-imidazole-4-methanol, 1-methyl, 1-methyl-1h-imidazol-4-yl-methanol
InChI Key NLEAEGDBKRBJAP-UHFFFAOYSA-N
SMILES CN1C=NC(CO)=C1
IUPAC Name (1-methyl-1H-imidazol-4-yl)methanol
Molecular Weight (g/mol) 112.13
PubChem CID 7060524
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methanol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.