Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-1H-imidazole-2-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 1-methyl-1H-imidazole-2-carboxylic acid |
Formula Weight | 126.11 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1H-imidazole-2-carboxylic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction