Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. M24511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24511G 1 g
M24515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24511G Supplier TCI America Supplier No. M24511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16681-77-9
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00129972
InChI Key OMAFFHIGWTVZOH-UHFFFAOYSA-N
PubChem CID 140123
IUPAC Name 1-methyltetrazole
SMILES CN1C=NN=N1

Specifications

Melting Point 40°C
Boiling Point 81°C
Quantity 1 g
UN Number 1325
Formula Weight 84.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.