1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar 437415Y

Quantity 4 x 100mL

 View more versions of this product

Catalog No. AA437415Y

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 872-50-4
    Molecular Formula C5H9NO
    Formula Weight 99.13
    MDL Number MFCD00003193
    Beilstein 106420
    Merck Index 14,6117
    Chemical Name or Material 1-Methyl-2-Pyrrolidinone, Anhydrous

    Specifications

    Formula Weight 99.13
    Quantity 4 x 100mL
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00003193
    Beilstein 106420
    Merck Index 14,6117
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360-H315-H319-H335-H227
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye
    Storage Note 1
    • Packaged under Argon
    RTECS Number UY5790000
    Molecular Formula C5H9NO
    CAS 872-50-4
    Chemical Name or Material 1-Methyl-2-Pyrrolidinone, Anhydrous
    Assay 99.5%

    GHS H Statement
    H360-H315-H319-H335-H227
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit