Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-n-octylimidazolium Bromide 98.0+%, TCI America™
SDP

Catalog No. M190425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M190425G 25 g
M19045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M190425G Supplier TCI America Supplier No. M190425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61545-99-1
Molecular Formula C12H23BrN2
Molecular Weight (g/mol) 275.234
InChI Key URVSXZLUUCVGQM-UHFFFAOYSA-M
PubChem CID 10849985
IUPAC Name 1-methyl-3-octylimidazol-1-ium;bromide
SMILES CCCCCCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 275.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.