Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-n-octylimidazolium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. M273225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M273225G 25 g
M27325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M273225G Supplier TCI America Supplier No. M273225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 244193-52-0
Molecular Formula C12H23BF4N2
Molecular Weight (g/mol) 282.13
MDL Number MFCD03095434
InChI Key GXZCAMSPWNHTAE-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium tetrafluoroborate, 1-methyl-3-n-octylimidazolium tetrafluoroborate, 3-methyl-1-octylimidazolium tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, omimbf4, dsstox_cid_27919, dsstox_rid_82672, dsstox_gsid_47943, 3-methyl-1-octylimidazoliumtetrafluoroborate, 1-methyl-3-octylimidazolium tetrafluoroborate hplc
PubChem CID 11460221
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Quantity 25 g
Formula Weight 282.13
Percent Purity ≥97.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.