Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-propylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. M303525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M303525G 25 g
M30355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M303525G Supplier TCI America Supplier No. M303525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79917-89-8
Molecular Formula C7H15ClN2
Molecular Weight (g/mol) 162.661
MDL Number MFCD17014728
InChI Key GYTJXQRCNBRFGU-UHFFFAOYSA-N
PubChem CID 67523443
IUPAC Name 1-methyl-3-propyl-1,2-dihydroimidazol-1-ium;chloride
SMILES CCCN1C[NH+](C=C1)C.[Cl-]

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 25 g
Formula Weight 160.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Methyl-3-propylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.