Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-iodo-1H-pyrazole, 97%

Catalog No. p-3736933
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 39806-90-1 | C4H5IN2 | 208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39806-90-1
MDL Number MFCD05663860
InChI Key RSDRDHPLXWMTRJ-UHFFFAOYSA-N
Synonym 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole
PubChem CID 2734773
IUPAC Name 4-iodo-1-methylpyrazole
SMILES CN1C=C(C=N1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 67.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5IN2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 208
Formula Weight 208
Percent Purity 97%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material 1-Methyl-4-iodo-1H-pyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.