Learn More
1-Methylcyclohexanecarboxylic acid, 99%
Description
1-Methyl-1-cyclohexanecarboxylic acid was used as internal standard during the determination of valproic acid metabolites. Also been used as an anticonvulsant drug and causes maturation of murine neuroblastoma cells in vitro. Used in the synthesis of p38 map kinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 36°C to 39°C |
Boiling Point | 236°C |
Flash Point | 100°C (212°F) |
Quantity | 25 g |
Beilstein | 1859220 |
Solubility Information | Soluble in chloroform and methanol. |
Formula Weight | 142.2 |
Percent Purity | 99% |
Chemical Name or Material | 1-Methylcyclohexanecarboxylic acid |
Safety and Handling
EINECSNumber | 214-371-3 |
RTECSNumber | GU8585500 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.