Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl methacrylate, 99%, stabilized

Catalog No. AC127140250
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Catalog No. AC127140250

Supplier: Thermo Scientific Chemicals 127140250

Only null left
Add to Cart
This item is not returnable. View return policy

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L | CAS: 80-62-6 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-62-6, 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Acidity 0.005 % max. (as methacrylic acid)
Melting Point -48.0°C
Color White to Yellow
Density 0.9300g/mL
Boiling Point 100.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH3
Refractive Index 1.4130 to 1.4150
Quantity 25 mL
Beilstein 02, II, 398
Specific Gravity 0.93
Merck Index 15, 6013
Solubility Information Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone
Stabilizer 10 to 110ppm MEHQ
Viscosity 0.6 mPa.s (20°C)
Formula Weight 100.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl methacrylate, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-297-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.