Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylhomopiperazine, 97%, Thermo Scientific™

Catalog No. p-7036950
Click to view available options
Quantity:
5 mL
25 mL
100 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4318-37-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059810
InChI Key FXHRAKUEZPSMLJ-UHFFFAOYSA-N
Synonym n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
PubChem CID 228349
IUPAC Name 1-methyl-1,4-diazepane
SMILES CN1CCCNCC1

Specifications

Density 0.918
Boiling Point 74°C to 75°C (35 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.477
Quantity 5 mL
UN Number UN2920
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material 1-Methylhomopiperazine
Hazard Category H226-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.