Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylpiperidine, 99%

Catalog No. AC127510025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 626-58-4 | C6H13N | 99.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-58-4
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00006005
InChI Key UZOFELREXGAFOI-UHFFFAOYSA-N
Synonym 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline
PubChem CID 69381
IUPAC Name 4-methylpiperidine
SMILES CC1CCNCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 4.0°C to 5.0°C
Color Yellow
Density 0.8400g/mL
Boiling Point 122.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Refractive Index 1.4448 to 1.4468
Quantity 2.5 kg
UN Number 2733
Specific Gravity 0.84
Solubility Information Solubility in water: miscible.
Formula Weight 99.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylpiperidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 210-954-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.