Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylnaphthalene, 96%

Click to view available options
Quantity:
100g
500g
90-12-0
C11H10
142.201
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene,naphthalene, 1-methyl,methylnaphthalene,naphthalene, methyl,polymethylnaphthalene,1-methyl-naphthalene,alpha-methyl naphthalenes,methyl naphthalene,1-methyl naphthalene,fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12
90-12-0
C11H10
142.201
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene,naphthalene, 1-methyl,methylnaphthalene,naphthalene, methyl,polymethylnaphthalene,1-methyl-naphthalene,alpha-methyl naphthalenes,methyl naphthalene,1-methyl naphthalene,fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12
Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

Solubility
Miscible with water, alcohol, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
Refractive Index 1.615
Density 1.024
Quantity 500g
Flash Point 82°C (179°F)
Melting Point -22°C
Boiling Point 240°C to 243°C
UN Number UN3082
Beilstein 506793
Solubility Information Miscible with water,alcohol,ethanol,ether and benzene.
Formula Weight 142.2
Percent Purity 96%
Chemical Name or Material 1-Methylnaphthalene
Show More Show Less

GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 201-966-8

RTECSNumber : QJ9630000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen