Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Methylstyrene, 97%, stabilized

Catalog No. AC127705000
Click to view available options
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 622-97-9 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-97-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008621
InChI Key JLBJTVDPSNHSKJ-UHFFFAOYSA-N
Synonym 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
PubChem CID 12161
IUPAC Name 1-ethenyl-4-methylbenzene
SMILES CC1=CC=C(C=C)C=C1

Specifications

Melting Point -34.0°C
Color Colorless to Yellow
Density 0.8960g/mL
Boiling Point 170.0°C to 175.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH=CH2
Refractive Index 1.5410 to 1.5430
Quantity 500 mL
Beilstein 05,485
Specific Gravity 0.896
Solubility Information Solubility in water: insoluble
Stabilizer 0.1% 3, 5-di-tert.-butylcatechol
Formula Weight 118.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Methylstyrene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause genetic defects.
Toxic to aquatic life with long lasting effects.
Ma
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 210-762-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.