Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylxanthine 97.0+%, TCI America™
SDP

Catalog No. M2432200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2432200MG 200 mg
M243250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2432200MG Supplier TCI America Supplier No. M2432200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6136-37-4
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00005561
InChI Key MVOYJPOZRLFTCP-UHFFFAOYSA-N
PubChem CID 80220
ChEBI CHEBI:68444
IUPAC Name 1-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C(=O)NC2=C(NC=N2)C1=O

Specifications

Quantity 200 mg
Formula Weight 166.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylxanthine
RTECSNumber ZD8575000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.