Learn More
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
Description
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 8°C to 10°C |
| Density | 1.365 |
| Boiling Point | >350°C |
| Refractive Index | 1.4115 |
| Quantity | 25 g |
| Sensitivity | Hygroscopic |
| Merck Index | 14,1581 |
| Solubility Information | Miscible with dichloromethane,chloroform,ethyl acetate. Immiscible with water,diethyl ether and hexane. |
| Formula Weight | 284.19 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.