Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Butylpyridinium chloride, 98%

Catalog No. AA1974009
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 1124-64-7 | C9H14ClN | 171.668 g/mol

The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-64-7
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD00060047
InChI Key POKOASTYJWUQJG-UHFFFAOYSA-M
Synonym 1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
PubChem CID 2734171
IUPAC Name 1-butylpyridin-1-ium;chloride
SMILES CCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Melting Point 131°C to 132°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 171.67
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-n-Butylpyridinium chloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.