Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthaldehyde, 95%

Catalog No. AC128095000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 66-77-3 | C11H8O | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-77-3
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004003
InChI Key SQAINHDHICKHLX-UHFFFAOYSA-N
Synonym 1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde
PubChem CID 6195
ChEBI CHEBI:52367
IUPAC Name naphthalene-1-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 1.0°C to 2.0°C
Color Brown-Yellow to Yellow
Density 1.1500g/mL
Boiling Point 160.0°C to 161.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6500 to 1.6540
Linear Formula C10H7CHO
Quantity 500 g
Beilstein 07, 400
Specific Gravity 1.15
Solubility Information Solubility in water: <1g/L (20°C)
Formula Weight 156.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Naphthaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-633-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.