Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthohydroxamic Acid 98.0+%, TCI America™
SDP

Catalog No. n00231g
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 6953-61-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00059546
InChI Key JRZGPWOEHDOVMC-UHFFFAOYSA-N
Synonym N-Hydroxynaphthalene-1-carboxamide
PubChem CID 23382
IUPAC Name N-hydroxynaphthalene-1-carboxamide
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)NO

Specifications

Quantity 1 g
Formula Weight 187.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthohydroxamic Acid
RTECSNumber QJ1894500
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.