Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol, 99%

Catalog No. p-7081749
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 90-15-3 | C10H8O | 144.173 g/mol

1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

Solubility
Soluble in benzene, chloroform, ether and ethanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00003930
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
PubChem CID 7005
ChEBI CHEBI:10319
IUPAC Name naphthalen-1-ol
SMILES C1=CC=C2C(=C1)C=CC=C2O

Specifications

Melting Point 95°C to 97°C
Density 1.28
Boiling Point 278°C to 280°C
Flash Point 125°C (257°F)
Odor Weak, Phenol-like
Quantity 100 g
UN Number UN2811
Beilstein 1817321
Sensitivity Air and light sensitive
Merck Index 14,6383
Solubility Information Soluble in benzene,chloroform,ether and ethanol.
Formula Weight 144.17
Percent Purity 99%
Chemical Name or Material 1-Naphthol
Show More Show Less
Hazard Category H302+H312-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H312-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-969-4
RTECSNumber QL2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.