Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
90-15-3
C10H8O
144.17
MFCD00003930
KJCVRFUGPWSIIH-UHFFFAOYSA-N
1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern
7005
CHEBI:10319
naphthalen-1-ol
C1=CC=C2C(=C1)C=CC=C2O
90-15-3
C10H8O
144.17
MFCD00003930
KJCVRFUGPWSIIH-UHFFFAOYSA-N
1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern
7005
CHEBI:10319
naphthalen-1-ol
C1=CC=C2C(=C1)C=CC=C2O
Boiling Point 278.0°C to 280.0°C
Color Beige-Gray to Brown
Quantity 500g
Infrared Spectrum Authentic
Melting Point 95.0°C to 97.0°C
Packaging Glass bottle
Flash Point 125°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C10H7OH
Beilstein 06, 596
Merck Index 15, 6468
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in benzene,alcohol and ether,freely soluble in chloroform,alkali hydroxide,solutions
Formula Weight 144.17
Physical Form Crystalline Flakes
Percent Purity ≥99%
Chemical Name or Material 1-Naphthol, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-969-4

RTECSNumber : QL2800000

TSCA : TSCA