Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthoxyacetic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
2976-75-2
C12H10O3
202.209
MFCD00003927
GHRYSOFWKRRLMI-UHFFFAOYSA-N
1-naphthoxyacetic acid,1-naphthyloxy acetic acid,2-naphthalen-1-yloxy acetic acid,1-naphthyloxyacetic acid,naphthyloxyacetic acid,2-1-naphthyloxy acetic acid,unii-9qi2464poa,1-naphthoxy acetic acid,naphthalen-1-yloxy acetic acid,acetic acid, 1-naphthalenyloxy
76313
CHEBI:44588
2-naphthalen-1-yloxyacetic acid
C1=CC=C2C(=C1)C=CC=C2OCC(=O)O
2976-75-2
C12H10O3
202.209
MFCD00003927
GHRYSOFWKRRLMI-UHFFFAOYSA-N
1-naphthoxyacetic acid,1-naphthyloxy acetic acid,2-naphthalen-1-yloxy acetic acid,1-naphthyloxyacetic acid,naphthyloxyacetic acid,2-1-naphthyloxy acetic acid,unii-9qi2464poa,1-naphthoxy acetic acid,naphthalen-1-yloxy acetic acid,acetic acid, 1-naphthalenyloxy
76313
CHEBI:44588
2-naphthalen-1-yloxyacetic acid
C1=CC=C2C(=C1)C=CC=C2OCC(=O)O
Packaging Glass bottle
Melting Point 193°C to 198°C
Quantity 25g
Infrared Spectrum Authentic
Color White to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C10H7OCH2COOH
Formula Weight 202.21
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1-Naphthoxyacetic acid, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 221-023-4

TSCA : TSCA