Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthoyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. N004925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N004925G 25 g
N00495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N004925G Supplier TCI America Supplier No. N004925G

Chemical Identifiers

CAS 14271-86-4
Molecular Formula C12H7NO
Molecular Weight (g/mol) 181.19
MDL Number MFCD00060239
InChI Key RITADAJGEQPFIR-UHFFFAOYSA-N
Synonym 1-Naphthalenecarbonyl Cyanide
PubChem CID 602423
IUPAC Name naphthalene-1-carbonyl cyanide
SMILES O=C(C#N)C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 100°C
Color Yellow
Boiling Point 182°C
Quantity 25 g
UN Number 3439
Formula Weight 181.19
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthoyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.