Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl butyrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3121-70-8
C14H14O2
214.26
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate,.alpha.-naphthyl butyrate,alpha-naphthyl butyrate,naphthalen-1-yl butyrate,butanoic acid, 1-naphthalenyl ester,butyric acid, 1-naphthyl ester,1-naphthylbutyrate,unii-9uzl9u0cik,butyric acid 1-naphthyl ester,9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21
3121-70-8
C14H14O2
214.26
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate,.alpha.-naphthyl butyrate,alpha-naphthyl butyrate,naphthalen-1-yl butyrate,butanoic acid, 1-naphthalenyl ester,butyric acid, 1-naphthyl ester,1-naphthylbutyrate,unii-9uzl9u0cik,butyric acid 1-naphthyl ester,9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21
Boiling Point 125°C to 130°C (1.0mmHg)
Flash Point 62°C
Specific Gravity 1.09
Density 1.0900g/mL
Quantity 1g
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: immiscible in water
Formula Weight 214.26
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Naphthyl butyrate, 98%
Show More Show Less

EINECSNumber : 221-500-7

TSCA : TSCA