Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 1-Naphthyl PP1
GREENER_CHOICE

Catalog No. 306310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-631-0 10 mg
30-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-631-0 Supplier Tocris Bioscience™ Supplier No. 3063/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Src family kinase inhibitor; also inhibits c-Abl

Selective inhibitor of src family kinases v-Src and c-Fyn as well as the tyrosine kinase c-Abl. (IC50 values are 1.0, 0.6, 0.6, 18 and 22 μM for v-Src, c-Fyn, c-Abl, CDK2 and CAMK II respectively). Preferentially inhibits mutant over wild-type kinases (IC50 values are 1.5 vs 1000 nM for I338G v-src and v-src respectively).

Chemical Identifiers

CAS 221243-82-9
Molecular Formula C19H19N5

Specifications

Quantity 10 mg
Formula Weight 317.39
Purity >99%
Molecular Weight (g/mol) 317.40
Chemical Name or Material 1-Naphthyl PP1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.