Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Naphthylamine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
Description
Applied as a synthesis reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red to White |
| Quantity | 10 g |
| UN Number | UN2811 |
| Sensitivity | Air and light sensitive |
| Merck Index | 14,6398 |
| Solubility Information | Soluble in water, alcohol, ether |
| Formula Weight | 179.65 |
| Percent Purity | 98% |
| Physical Form | Crystalline |
| Chemical Name or Material | 1-Naphthylamine hydrochloride |
Safety and Handling
| Hazard Category | H302-H350 |
| Hazard Statement | GHS H Statement H350-H302 May cause cancer. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 209-014-3 |
| RTECSNumber | QM3150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction