Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Naphthylamine 99.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Melting Point | 50°C |
Color | Red-White |
Boiling Point | 301°C |
Quantity | 100 g |
UN Number | 2077 |
Formula Weight | 143.19 |
Percent Purity | ≥99.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 1-Naphthylamine |
Safety and Handling
EINECSNumber | (4)-0321 |
RTECSNumber | QM1400000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction