Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Naphthylmethyl)boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. aah62764md
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(1-Naphthylmethyl)boronic acid pinacol ester is employed as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475250-57-8
Molecular Formula C17H21BO2
Molecular Weight (g/mol) 268.163
MDL Number MFCD22419278
InChI Key VYMBZQMGIGIALJ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-naphthalen-1-ylmethyl-1,3,2-dioxaborolane, naphthalen-1-ylmethyl boronic acid pinacol ester, 2-1-naphthylmethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl methyl naphthalene
PubChem CID 11254165
IUPAC Name 4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC3=CC=CC=C32

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 268.16
Percent Purity 96%
Chemical Name or Material (1-Naphthylmethyl)boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.