Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitropyrene 98.0+%, TCI America™
SDP

Catalog No. N041925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N041925G 25 g
N04195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N041925G Supplier TCI America Supplier No. N041925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5522-43-0
Molecular Formula C16H9NO2
Molecular Weight (g/mol) 247.25
MDL Number MFCD00004138
InChI Key ALRLPDGCPYIVHP-UHFFFAOYSA-N
Synonym 3-nitropyrene, pyrene, 1-nitro, pyrene, nitro, nitropyrene, ccris 455, 1-nitro-pyrene, unii-td1665i8q4, nitroarenes, polycyclic aromatic compounds, 1-nitropyrene
PubChem CID 21694
ChEBI CHEBI:34107
IUPAC Name 1-nitropyrene
SMILES [O-][N+](=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 152°C
Color Yellow
Quantity 25 g
Formula Weight 247.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitropyrene
EINECSNumber (4)-0391
RTECSNumber UR2480000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.