Alfa Aesar™ 1-Nonyne, 98%

Manufacturer: Alfa Aesar™ B2426306

Quantity 5g

 View more versions of this product

Catalog No. AAB2426306

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 3452-09-3
    Molecular Formula C9H16
    Linear Formula CH3(CH2)6C≡CH
    Formula Weight 124.23
    MDL Number MFCD00009567
    UN Number UN3295
    Beilstein 1736573
    Synonym n-Heptylacetylene
    Chemical Name or Material 1-Nonyne

    Specifications

    Formula Weight 124.23
    TSCA TSCA
    Molecular Formula C9H16
    Linear Formula CH3(CH2)6C≡CH
    MDL Number MFCD00009567
    CAS 3452-09-3
    UN Number UN3295
    Beilstein 1736573
    Synonym n-Heptylacetylene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P301+P330+P331-P315
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Flash Point 33°C (91°F)
    Odor Petrol-like
    Boiling Point 149° to 151°C
    Melting Point −50°C
    Quantity 5g
    Chemical Name or Material 1-Nonyne
    Assay 98%

    Description

    Applications
    1-Nonyne has been used in enantioselective synthesis of cladospolide B, C and (ent)-cladospolide D.

    Solubility
    Insoluble in water.

    Notes
    Store away from oxidizing agents, heavy metal powders.

    GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapour.

    GHS P Statement
    P301+P330+P331-P315
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit