Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 98%

Catalog No. p-7046432
Click to view available options
Quantity:
1 g
5 g

CAS: 6974-32-9 | C28H24O9 | 504.491 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6974-32-9
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.491
MDL Number MFCD00005357
InChI Key GCZABPLTDYVJMP-CBUXHAPBSA-N
Synonym 1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
PubChem CID 81455
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
Beilstein 100243
Optical Rotation +44° (c=0.5 in Chloroform)
Formula Weight 504.5
Percent Purity 98%
Chemical Name or Material 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
EINECSNumber 230-220-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.