Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(o-Tolyl)piperazine dihydrochloride, 98%, Thermo Scientific™

Catalog No. AAB2260414
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2260414 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2260414 Supplier Thermo Scientific Chemicals Supplier No. B2260414

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 70849-60-4
Molecular Formula C11H17ClN2
Quantity 25g
MDL Number MFCD00039030
Beilstein 4019516
Sensitivity Hygroscopic
Synonym 1-o-tolyl piperazine hydrochloride, n-2-methylphenyl piperazine hydrochloride, 1-2-methylphenyl piperazine hydrochloride, 1-2-tolyl piperazine hydrochloride, piperazine, 1-2-methylphenyl-, monohydrochloride, piperazine, 1-2-methylphenyl-, hydrochloride, acmc-209ryj, 1-o-tolyl piperazinehydrochloride, 1-o-tolyl-piperazine hydrochloride, n-o-tolyl piperazine hydrochloride
InChI Key LZNYIWJDVYCKDU-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1N2CCNCC2.Cl
IUPAC Name 1-(2-methylphenyl)piperazine;hydrochloride
Molecular Weight (g/mol) 212.721
PubChem CID 12227895
Formula Weight 249.19
Percent Purity 98%
Chemical Name or Material 1-(o-Tolyl)piperazine dihydrochloride
Show More Show Less
Hazard Category H302-H311-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.