Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Octadecene

Catalog No. O8061L Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 mL
1 L
Catalog No. Quantity
O8061L 1 L
O80625ML 25 mL
2 options

Catalog No. O8061L

Supplier: Sigma Aldrich O8061L

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octadecene

Specifications

CAS 112-88-9
Melting Point 14°C to 16°C (lit.)
Density 0.789 g/mL (at 25°C (literature))
Boiling Point 314.4°C ± 5.0°C (760 mmHg), 179°C (15 mmHg), 314.4°C ± 5.0°C (760 mmHg)
Molecular Formula C18H36
Refractive Index n20/D 1.444 (literature)
Linear Formula CH3(CH2)15 CH=CH2
MDL Number MFCD00009003
Quantity 1 L
Synonym ODE
Molecular Weight (g/mol) 252.48
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.