Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals

Catalog No. AC416360250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416360250 25 g
AC416361000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416360250 Supplier Thermo Scientific Chemicals Supplier No. 416360250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Absorbance (2.7 g/50 ml water) (acidified with ACS grade Phosphoric Acid to pH 3), 0.04 max. at 254nm, 0.04 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
Melting Point >300°C
Color White
Loss on Drying 1.5% max. (1 g, 110°C, 75 min)
Infrared Spectrum Authentic
Linear Formula CH3(CH2)7SO3Na
Quantity 25 g
Beilstein 04, III, 25
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
Formula Weight 216.28
Grade HPLC
Physical Form Crystalline Solid
Chemical Name or Material 1-Octanesulfonic Acid, sodium salt
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-195-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.