Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanol, 99%

Catalog No. p-7022968
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL

CAS: 111-87-5 | C8H18O | 130.23 g/mol

1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point °C to 16°C
Density 0.825
Boiling Point 193°C to 195°C
Flash Point 81°C (177°F)
Assay Percent Range 99%
Refractive Index 1.429
Linear Formula CH3(CH2)7OH
Quantity 500 mL
Beilstein 1697461
Merck Index 14,6751
Solubility Information Miscible with ethanol,ether and propylene glycol. Immiscible with water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material 1-Octanol
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H315-H319-H227
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 203-917-6
RTECSNumber RH6550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.