Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanol 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal Drum
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol,octanol,octyl alcohol,n-octanol,capryl alcohol,caprylic alcohol,n-octyl alcohol,heptyl carbinol,1-hydroxyoctane,primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol,octanol,octyl alcohol,n-octanol,capryl alcohol,caprylic alcohol,n-octyl alcohol,heptyl carbinol,1-hydroxyoctane,primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO
Viscosity 9 mPa.s (20°C)
Color Colorless
Infrared Spectrum Authentic
Quantity 1L
Melting Point -16.0°C
Boiling Point 195.0°C
Density 0.8240g/mL
Flash Point 81°C
Packaging Glass bottle
Refractive Index 1.4280 to 1.4310
Specific Gravity 0.824
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)7OH
Beilstein 01, 418
Merck Index 15, 6840
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,and mineral oil,soluble in propylene glycol,insoluble in glycerol
Formula Weight 130.23
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material 1-Octanol
Show More Show Less

GHS H StatementCauses serious eye irritation.Harmful to aquatic life with long lasting effects.

GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.Avoid release to the environment.

;GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing.Avoid release to the env