Learn More
1-Octanol, natural, 98%
Description
1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | -16°C |
| Color | Colorless |
| Density | 0.825 g/mL |
| Boiling Point | 193°C to 195°C |
| Flash Point | 81°C (178°F) |
| Odor | Sweet |
| Refractive Index | 1.4295 |
| Quantity | 10,000 mL |
| Solubility Information | Not miscible in water. |
| Formula Weight | 130.23 |
| Show More |
Safety and Handling
| Hazard Category | H227-H319 |
| Hazard Statement | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
| Precautionary Statement | P210-P264b-P280-P305+P351+P338-P370+P378q-P501c |
| EINECSNumber | 203-917-6 |
| RTECSNumber | RH6550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.