Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. T29301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29301G 1 g
T29305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29301G Supplier TCI America Supplier No. T29301G

Chemical Identifiers

CAS 77451-51-5
Molecular Formula C9H8N4O4S
Molecular Weight (g/mol) 268.25
MDL Number MFCD00009756
InChI Key ZQMJAWSQRGYFBM-UHFFFAOYSA-N
Synonym 3-nitro-1-tosyl-1h-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-methylphenyl sulfonyl-3-nitro-1,2,4-triazole, 1-4-methylbenzenesulfonyl-3-nitro-1,2,4-triazole, tsnt, 3-nitro-1-tosyl-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazol, 3-nitro-1-p-tolylsulfonyl-1,2,4-triazole, 3-nitro-1-p-toluenesulfonyl-1,2,4-triazole
PubChem CID 688072
IUPAC Name 1-(4-methylbenzenesulfonyl)-3-nitro-1H-1,2,4-triazole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 268.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.