Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentadecene, 97%

Catalog No. p-7035789
Click to view available options
Quantity:
5 g
25 g

CAS: 13360-61-7 | C15H30 | 210.405 g/mol

1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13360-61-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00008987
InChI Key PJLHTVIBELQURV-UHFFFAOYSA-N
Synonym 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q
PubChem CID 25913
ChEBI CHEBI:77506
IUPAC Name pentadec-1-ene
SMILES CCCCCCCCCCCCCC=C

Specifications

Melting Point -4°C
Density 0.775
Boiling Point 268°C to 269°C
Flash Point 110°C (230°F)
Refractive Index 1.439
Linear Formula CH3(CH2)12CH=CH2
Quantity 5 g
Beilstein 1751011
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 210.4
Percent Purity 97%
Chemical Name or Material 1-Pentadecene
Show More Show Less
EINECSNumber 236-414-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.