Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentadecene 95.0+%, TCI America™
SDP

Catalog No. P062725ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 13360-61-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00008987
InChI Key PJLHTVIBELQURV-UHFFFAOYSA-N
Synonym 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q
PubChem CID 25913
ChEBI CHEBI:77506
IUPAC Name pentadec-1-ene
SMILES CCCCCCCCCCCCCC=C

Specifications

Melting Point -4°C
Color Colorless
Boiling Point 268°C
Quantity 25 mL
Formula Weight 210.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentadecene
EINECSNumber (2)-0027
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.